Oxalic acid bis(cyclohexylidenehydrazide), 98%
Oxalic acid bis(cyclohexylidenehydrazide), 98%
Oxalic acid bis(cyclohexylidenehydrazide), 98%
Oxalic acid bis(cyclohexylidenehydrazide), 98%
Thermo Scientific Chemicals
Oxalic acid bis(cyclohexylidenehydrazide), 98%
CAS: 370-81-0 | C14H22N4O2 | 278.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA154420250
Price (MYR)
453.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
453.00
BOT
Add to cart
Specifications
Chemical Name or MaterialOxalic acid bis(cyclohexylidenehydrazide)
Name Note98%
CAS370-81-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: