n-Butyrophenone, 99%
n-Butyrophenone, 99%
n-Butyrophenone, 99%
n-Butyrophenone, 99%
Thermo Scientific Chemicals
n-Butyrophenone, 99%
CAS: 495-40-9 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA154411000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Butyrophenone
CAS495-40-9
Melting Point11.0°C to 13.0°C
ColorColorless to Yellow
Density0.9880g/mL