5-Bromoisatin, 98%
5-Bromoisatin, 98%
5-Bromoisatin, 98%
Thermo Scientific Chemicals
5-Bromoisatin, 98%
CAS: 87-48-9 | C8H4BrNO2 | 226.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Plastic Bottle
Catalog number FSA154230250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or Material5-Bromoisatin
CAS87-48-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0