D(+)-Malic acid, 98%
D(+)-Malic acid, 98%
D(+)-Malic acid, 98%
D(+)-Malic acid, 98%
Thermo Scientific Chemicals
D(+)-Malic acid, 98%
CAS: 636-61-3 | C4H6O5 | 134.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA153500010
Price (MYR)
272.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
272.00
EA
Add to cart
Specifications
Chemical Name or MaterialD(+)-Malic acid
CAS636-61-3
Melting Point99.0°C to 104.0°C
Health Hazard 1Danger