Fast Garnet GBC Base 97%, Thermo Scientific Chemicals
Fast Garnet GBC Base 97%, Thermo Scientific Chemicals
Fast Garnet GBC Base 97%, Thermo Scientific Chemicals
Fast Garnet GBC Base 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fast Garnet GBC Base 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA153260250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFast Garnet GBC base
Name Note97%
CAS97-56-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May cause an allergic skin reaction.
May cause cancer.