Rhodanine, 99%
Rhodanine, 99%
Rhodanine, 99%
Thermo Scientific Chemicals
Rhodanine, 99%
CAS: 141-84-4 | C3H3NOS2 | 133.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA152601000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodanine
Melting Point167°C to 170°C
Name Note99%
CAS141-84-4
Health Hazard 2GHS H Statement: