Phenylacetylene, 98%, pure
Phenylacetylene, 98%, pure
Phenylacetylene, 98%, pure
Phenylacetylene, 98%, pure
Thermo Scientific Chemicals
Phenylacetylene, 98%, pure
CAS: 536-74-3 | C8H6 | 102.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA152461000
Price (MYR)
856.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
856.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenylacetylene
CAS536-74-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.