1,2-Diaminopropane, 99%
1,2-Diaminopropane, 99%
1,2-Diaminopropane, 99%
1,2-Diaminopropane, 99%
Thermo Scientific Chemicals
1,2-Diaminopropane, 99%
CAS: 78-90-0 | C3H10N2 | 74.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA152402500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Diaminopropane
CAS78-90-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0