Trichloroacetic acid, 99%, extra pure
Trichloroacetic acid, 99%, extra pure
Trichloroacetic acid, 99%, extra pure
Trichloroacetic acid, 99%, extra pure
Thermo Scientific Chemicals
Trichloroacetic acid, 99%, extra pure
CAS: 76-03-9 | C2HCl3O2 | 163.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA152135000
Price (MYR)
629.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
629.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrichloroacetic acid
Name NoteExtra Pure
CAS76-03-9
Melting Point52.0°C to 58.0°C
Boiling Point196.0°C