1,1'-Carbonyldiimidazole, 97%
1,1'-Carbonyldiimidazole, 97%
1,1'-Carbonyldiimidazole, 97%
1,1'-Carbonyldiimidazole, 97%
Thermo Scientific Chemicals
1,1'-Carbonyldiimidazole, 97%
CAS: 530-62-1 | C7H6N4O | 162.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA151815000
Price (MYR)
5,284.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,284.00
EA
Add to cart
Specifications
Chemical Name or Material1,1′-Carbonyldiimidazole
CAS530-62-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Coupling reagent
RUO – Research Use Only