Phenylhydrazine Hydrochloride, 99+%
Phenylhydrazine Hydrochloride, 99+%
Phenylhydrazine Hydrochloride, 99+%
Phenylhydrazine Hydrochloride, 99+%
Thermo Scientific Chemicals
Phenylhydrazine Hydrochloride, 99+%
CAS: 59-88-1 | C6H9ClN2 | 144.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA151555000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenylhydrazine hydrochloride
Name Note99+%
CAS59-88-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Suspected of causing genetic defects. May cause cancer. May cause an allergic skin reaction. Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life.