4-(4-Nitrophenylazo)resorcinol, 90%
4-(4-Nitrophenylazo)resorcinol, 90%
4-(4-Nitrophenylazo)resorcinol, 90%
4-(4-Nitrophenylazo)resorcinol, 90%
Thermo Scientific Chemicals
4-(4-Nitrophenylazo)resorcinol, 90%
CAS: 74-39-5 | C12H9N3O4 | 259.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA151511000
Price (MYR)
1,021.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,021.00
BOT
Add to cart
Specifications
Chemical Name or Material4-(4-Nitrophenylazo)resorcinol
Name Notedye content ca 90%
CAS74-39-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement