Isatin, 98%
Isatin, 98%
Isatin, 98%
Thermo Scientific Chemicals
Isatin, 98%
CAS: 91-56-5 | C8H5NO2 | 147.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA151491000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsatin
Name Note98%
CAS91-56-5
Melting Point201°C to 204°C
ColorOrange