3-Aminopropyltriethoxysilane, 99%
3-Aminopropyltriethoxysilane, 99%
3-Aminopropyltriethoxysilane, 99%
3-Aminopropyltriethoxysilane, 99%
Thermo Scientific Chemicals
3-Aminopropyltriethoxysilane, 99%
CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA151085000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Aminopropyltriethoxysilane
CAS919-30-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre