Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol
Thermo Scientific Chemicals
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol
CAS: 77-73-6 | C10H12 | 132.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA150760010
Price (MYR)
565.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
565.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDicyclopentadiene
CAS77-73-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Highly flammable
CAS98-29-3