Diphenylamine, 99%, pure
Diphenylamine, 99%, pure
Diphenylamine, 99%, pure
Diphenylamine, 99%, pure
Thermo Scientific Chemicals
Diphenylamine, 99%, pure
CAS: 122-39-4 | C12H11N | 169.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number FSA150741000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiphenylamine
CAS122-39-4
Health Hazard 1GHS Signal Word: Danger
Melting Point52.0°C to 54.0°C
ColorOrange to White
View more
Indicator
RUO – Research Use Only