Dimethyl succinate, 99%
Dimethyl succinate, 99%
Dimethyl succinate, 99%
Dimethyl succinate, 99%
Thermo Scientific Chemicals
Dimethyl succinate, 99%
CAS: 106-65-0 | C6H10O4 | 146.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA150735000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl succinate
Name Note99%
Acidity0.2mg KOH/g max.
CAS106-65-0
Health Hazard 1Warning