Histamine dihydrochloride, 99%
Histamine dihydrochloride, 99%
Histamine dihydrochloride, 99%
Histamine dihydrochloride, 99%
Thermo Scientific Chemicals
Histamine dihydrochloride, 99%
CAS: 56-92-8 | C5H11Cl2N3 | 184.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA150620250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHistamine dihydrochloride
CAS56-92-8
Health Hazard 1Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Respiratory sensitisation (category 1), Skin sensitisation (category 1), Skin corrosion/irritation (category 2)
Health Hazard 2Causes serious eye irritation, May cause respiratory irritation, May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction, Causes skin irritation
Melting Point249.0°C to 252.0°C