4-Chlorobenzaldehyde, 98%
4-Chlorobenzaldehyde, 98%
4-Chlorobenzaldehyde, 98%
4-Chlorobenzaldehyde, 98%
Thermo Scientific Chemicals
4-Chlorobenzaldehyde, 98%
CAS: 104-88-1 | C7H5ClO | 140.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA150442500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Chlorobenzaldehyde
CAS104-88-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN:
View more