Thymol, 99%
Thymol, 99%
Thymol, 99%
Thermo Scientific Chemicals
Thymol, 99%
CAS: 89-83-8 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA150335000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThymol
Name Note99%
CAS89-83-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic to aquatic life with long lasting effects.