4-Chlorobenzoyl chloride, 99+%
4-Chlorobenzoyl chloride, 99+%
4-Chlorobenzoyl chloride, 99+%
4-Chlorobenzoyl chloride, 99+%
Thermo Scientific Chemicals
4-Chlorobenzoyl chloride, 99+%
CAS: 122-01-0 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA150191000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Chlorobenzoyl chloride
CAS122-01-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat