DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
Thermo Scientific Chemicals
DL-3-Methyl-2-butanol, 98%
CAS: 598-75-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA149931000
Price (MYR)
465.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
465.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDL-3-Methyl-2-butanol
CAS598-75-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if inhaled.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfac