Acetic acid, sodium salt, 99+%, extra pure, anhydrous
Acetic acid, sodium salt, 99+%, extra pure, anhydrous
Acetic acid, sodium salt, 99+%, extra pure, anhydrous
Acetic acid, sodium salt, 99+%, extra pure, anhydrous
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, extra pure, anhydrous
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA149610010
Price (MYR)
464.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
464.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt
Name NoteAnhydrous
CAS127-09-3
Melting Point324.0°C
CAS Min %99.0