4-Methyl-2-pentanol, 99+%
4-Methyl-2-pentanol, 99+%
4-Methyl-2-pentanol, 99+%
4-Methyl-2-pentanol, 99+%
Thermo Scientific Chemicals
4-Methyl-2-pentanol, 99+%
CAS: 108-11-2 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA149385000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Methyl-2-pentanol
CAS108-11-2
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0