3,3'-Dithiodipropionic acid, 98%
3,3'-Dithiodipropionic acid, 98%
3,3'-Dithiodipropionic acid, 98%
3,3'-Dithiodipropionic acid, 98%
Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%
CAS: 1119-62-6 | C6H10O4S2 | 210.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA148902500
Price (MYR)
726.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
726.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 3'-Dithiodipropionic acid
Name Note98%
CAS1119-62-6
CAS Min %98
Health Hazard 1GHS Signal Word: Danger