2-Aminobenzothiazole, 97%
2-Aminobenzothiazole, 97%
2-Aminobenzothiazole, 97%
2-Aminobenzothiazole, 97%
Thermo Scientific Chemicals
2-Aminobenzothiazole, 97%
CAS: 136-95-8 | C7H6N2S | 150.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA148811000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Aminobenzothiazole
Name Note97%
CAS136-95-8
CAS Min %96.0
CAS Max %100.0