Phenyl acetate, 97%
Phenyl acetate, 97%
Phenyl acetate, 97%
Thermo Scientific Chemicals
Phenyl acetate, 97%
CAS: 122-79-2 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA148770050
Price (MYR)
196.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
196.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenyl acetate
Name Note97%
CAS122-79-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement