Acetaldehyde ammonia trimer, 98%
Acetaldehyde ammonia trimer, 98%
Acetaldehyde ammonia trimer, 98%
Acetaldehyde ammonia trimer, 98%
Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%
CAS: 58052-80-5 | C6H18N3 | 132.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA148535000
Price (MYR)
2,833.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,833.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetaldehyde ammonia trimer
Name Note98%
CAS58052-80-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning