Sodium formate, 99%
Sodium formate, 99%
Sodium formate, 99%
Thermo Scientific Chemicals
Sodium formate, 99%
CAS: 141-53-7 | CHNaO2 | 68.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA148230010
Price (MYR)
334.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
334.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium formate
CAS141-53-7
Melting Point258.0°C to 261.0°C
ColorWhite
Boiling Point360.0°C