2,4-Dichlorophenol, 99%
2,4-Dichlorophenol, 99%
2,4-Dichlorophenol, 99%
2,4-Dichlorophenol, 99%
Thermo Scientific Chemicals
2,4-Dichlorophenol, 99%
CAS: 120-83-2 | C6H4Cl2O | 163.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA147725000
Price (MYR)
569.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
569.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4-Dichlorophenol
Name Note99%
CAS120-83-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger