Octadecyltrichlorosilane, 95%
Octadecyltrichlorosilane, 95%
Octadecyltrichlorosilane, 95%
Octadecyltrichlorosilane, 95%
Thermo Scientific Chemicals
Octadecyltrichlorosilane, 95%
CAS: 112-04-9 | C18H37Cl3Si | 387.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA147405000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOctadecyltrichlorosilane
CAS112-04-9
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0