2-Chlorobenzoyl chloride, 98%
2-Chlorobenzoyl chloride, 98%
2-Chlorobenzoyl chloride, 98%
2-Chlorobenzoyl chloride, 98%
Thermo Scientific Chemicals
2-Chlorobenzoyl chloride, 98%
CAS: 609-65-4 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA147142500
Price (MYR)
455.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
455.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Chlorobenzoyl chloride
CAS609-65-4
ColorColorless to Yellow
Density1.38g/mL
Infrared SpectrumAuthentic