1,2,3,4-Tetrahydronaphthalene, 98+%, pure
1,2,3,4-Tetrahydronaphthalene, 98+%, pure
1,2,3,4-Tetrahydronaphthalene, 98+%, pure
1,2,3,4-Tetrahydronaphthalene, 98+%, pure
Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure
CAS: 119-64-2 | C10H12 | 132.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number FSA146730100
Price (MYR)
3,812.00
EA
Add to cart
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
3,812.00
EA
Add to cart
Specifications
Chemical Name or Material1,2,3,4-Tetrahydronaphthalene
CAS119-64-2
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0