Safranin O, 95%, pure, high purity biological stain
Safranin O, 95%, pure, high purity biological stain
Safranin O, 95%, pure, high purity biological stain
Safranin O, 95%, pure, high purity biological stain
Thermo Scientific Chemicals
Safranin O, 95%, pure, high purity biological stain
Safranine O, C20H19ClN4, CAS Number-477-73-6, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride, Basic Red 2, C.I. 50240, 100g, 95%, Brown or Brown-Red to Green, 94% min., 350.84, Pure, Warning, GHS H Statement: Causes serious eye irritation. Causes skin irritation. | CAS: 477-73-6 | C20H19ClN4 | 350.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA146640250
Price (MYR)
448.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
448.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSafranine O
Name NotePure
CAS477-73-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
View more
Indicator
RUO – Research Use Only