4-Heptanone, 98%
4-Heptanone, 98%
4-Heptanone, 98%
Thermo Scientific Chemicals
4-Heptanone, 98%
CAS: 123-19-3 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA146555000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Heptanone
Name Note98%
CAS123-19-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning