Yohimbine hydrochloride, 99%
Yohimbine hydrochloride, 99%
Yohimbine hydrochloride, 99%
Yohimbine hydrochloride, 99%
Thermo Scientific Chemicals
Yohimbine hydrochloride, 99%
Yohimbine hydrochloride, CAS # 65-19-0, is an α2-adrenergic receptor antagonist, which blocks the central α2-adrenegic receptors in the brain. | CAS: 65-19-0 | C21H27ClN2O3 | 390.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA141030050
Price (MYR)
692.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
692.00
BOT
Add to cart
Specifications
Chemical Name or MaterialYohimbine hydrochloride
CAS65-19-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
General Description
  • Yohimbine hydrochloride is an indole alkaloid derived from the tree bark
  • It is an antagonist of α2-adrenergic receptors in the brain, reducing the effects of xylazine, which is an α2-adrenergic agonist
Applications
  • Yohimbine hydrochloride blocks the central α2-adrenegic receptors in the brain
  • It can reduce or eradicate effects of xylazine, which is an α2-adrenergic agonist
  • It can have an antidiuretic effect
RUO – Research Use Only
General References:
  1. Zhang, W. and Roomans, G.M. A yohimbine-dependent, UK14,304 induced ion transient in HT29 cells studied by X-ray microanalysis. Scanning microscopy, 1996, 10 (1), 293-299.
  2. Susan, X. Jiang et. al. Reversible inhibition of intracellular calcium influx through NMDA receptors by imidazoline I2receptor antagonists. European Journal of Pharmacology. 2010, 629,(1), 12-19.