3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
Thermo Scientific Chemicals
3,4-Dimethoxybenzylamine, 97%
CAS: 5763-61-1 | C9H13NO2 | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA140830050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 4-Dimethoxybenzylamine
CAS5763-61-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0