L-Valine, 99%
L-Valine, 99%
L-Valine, 99%
Thermo Scientific Chemicals
L-Valine, 99%
CAS: 72-18-4 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic Bottle
Catalog number FSA140811000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Valine
Melting Point315.0°C
Name Note99%
Ammonium200ppm max.
CAS72-18-4