L(-)-Tryptophan, 99%
L(-)-Tryptophan, 99%
L(-)-Tryptophan, 99%
L(-)-Tryptophan, 99%
Thermo Scientific Chemicals
L(-)-Tryptophan, 99%
L(-)-Tryptophan, CAS # 73-22-3, is an essential amino acid necessary for the biosynthesis of proteins. | CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA140595000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL(-)-Tryptophan
Melting Point280.0°C to 285.0°C
Name Note99%
CAS73-22-3
ColorWhite to White-Yellow
View more
General Description
  • L(-)-Tryptophan is a nonpolar aromatic compound and essential amino acid required for protein synthesis
  • It is a biochemical precursor of niacin, indole alkaloids, and serotonin
Applications
  • L-Tryptophan acts as a natural dietary supplement and has laboratory applications in the research of antidepressants, anxiolytics, and sleep aids
  • It is a precursor of metabolites that are important in the laboratory study of mammalian physiology, including gastrointestinal functions, immunity, metabolism, and the nervous system
RUO – Research Use Only