Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Tris(hydroxymethyl)aminomethane, 99+%, C4H11NO3, CAS Number-77-86-1, 2-amino-2-hydroxymethyl propane-1,3-diol, tham, tris, trisamine, tris base, tris buffer, tris hydroxymethyl aminomethane, trizma, trometamol, tromethamine, 2.5kg | CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA140505000
Price (MYR)
517.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
517.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTris(hydroxymethyl)aminomethane
CAS77-86-1
Absorbance(40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
View more
Biological buffer
RUO – Research Use Only