Triphenylphosphine oxide, 99%
Triphenylphosphine oxide, 99%
Triphenylphosphine oxide, 99%
Triphenylphosphine oxide, 99%
Thermo Scientific Chemicals
Triphenylphosphine oxide, 99%
CAS: 791-28-6 | C18H15OP | 278.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA140431000
Price (MYR)
469.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
469.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriphenylphosphine oxide
CAS791-28-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent