Triphenylphosphine, 99%
Triphenylphosphine, 99%
Triphenylphosphine, 99%
Triphenylphosphine, 99%
Thermo Scientific Chemicals
Triphenylphosphine, 99%
CAS: 603-35-0 | C18H15P | 262.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA140422500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTriphenylphosphine
CAS603-35-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.