Tri-n-octylamine, 97%
Tri-n-octylamine, 97%
Tri-n-octylamine, 97%
Tri-n-octylamine, 97%
Thermo Scientific Chemicals
Tri-n-octylamine, 97%
CAS: 1116-76-3 | C24H51N | 353.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA140285000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-n-octylamine
CAS1116-76-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the envir