Trimethylacetyl chloride, 99%
Trimethylacetyl chloride, 99%
Trimethylacetyl chloride, 99%
Trimethylacetyl chloride, 99%
Thermo Scientific Chemicals
Trimethylacetyl chloride, 99%
CAS: 3282-30-2 | C5H9ClO | 120.58 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA140052500250 gGlass bottle
Catalog number FSA140052500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTrimethylacetyl chloride
Name Note99%
CAS3282-30-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Highly flammable liquid and vapor.