Triethyl phosphate, 99%
Triethyl phosphate, 99%
Triethyl phosphate, 99%
Triethyl phosphate, 99%
Thermo Scientific Chemicals
Triethyl phosphate, 99%
CAS: 78-40-0 | C6H15O4P | 182.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass bottle
Catalog number FSA139680025
Price (MYR)
1,159.00
BOT
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,159.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriethyl phosphate
Name Note99%
CAS78-40-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: