Triethyl orthoformate, 98%
Triethyl orthoformate, 98%
Triethyl orthoformate, 98%
Triethyl orthoformate, 98%
Thermo Scientific Chemicals
Triethyl orthoformate, 98%
CAS: 122-51-0 | C7H16O3 | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA139660010
Price (MYR)
469.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
469.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriethyl orthoformate
CAS122-51-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
View more
Alkylation reagent
RUO – Research Use Only