Triethylene glycol dimethyl ether, 99%, stabilized
Triethylene glycol dimethyl ether, 99%, stabilized
Triethylene glycol dimethyl ether, 99%, stabilized
Triethylene glycol dimethyl ether, 99%, stabilized
Thermo Scientific Chemicals
Triethylene glycol dimethyl ether, 99%, stabilized
CAS: 112-49-2 | C8H18O4 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA139600010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTriethylene glycol dimethyl ether
Name NoteStabilized
Acidity0.05mg KOH/g max.
CAS112-49-2
Health Hazard 1GHS Signal Word: Danger