n-Tridecane, 99+%
n-Tridecane, 99+%
n-Tridecane, 99+%
Thermo Scientific Chemicals
n-Tridecane, 99+%
CAS: 629-50-5 | C13H28 | 184.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA139515000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Tridecane
CAS629-50-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0