p-Toluenesulfonic acid monohydrate, 99%, extra pure
p-Toluenesulfonic acid monohydrate, 99%, extra pure
p-Toluenesulfonic acid monohydrate, 99%, extra pure
p-Toluenesulfonic acid monohydrate, 99%, extra pure
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA139025000
Price (MYR)
551.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
551.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Toluenesulfonic acid monohydrate
Name Note>98.5% (total TSA)
CAS6192-52-5
Melting Point100.0°C to 108.0°C
Health Hazard 1Danger