Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Thermo Scientific Chemicals
Tetramethylthiuram disulfide, 97%
CAS: 137-26-8 | C6H12N2S4 | 240.416 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA138485000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylthiuram disulfide
Name Note97%
CAS137-26-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause an allergic skin reaction.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.